Learn More
Chemglass Life Sciences Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 22 x 40mm, 1oz/Pkg, Approx 77/Oz
SDP
Catalog No. 501532237
Encompass_Preferred
Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 22 x 40mm, 1oz/Pkg, Approx 77/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1.5 thickness (0.16 to 0.19mm). Sold in packages of 1oz.
Catalog No. 50-153-2237 Supplier Chemglass Life Sciences Supplier No. CLS17642240
Cover Slip, Rectangular, #1.5 Corning 0211 Glass, 22 x 40mm, 1oz/Pkg, Approx 77/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1.5 thickness (0.16 to 0.19mm). Sold in packages of 1oz.