Learn More
Electron Microscopy Sciences Coverslip Tape 70 MT
SDP
Supplier:  Electron Microscopy Sciences 6400505
Encompass
Coverslip Tape has the same clarity as glass. The tape is loaded on a spool with 140 mm diameter core. This roll of coverslip tape eliminates the need for manual cover-slipping and extended user contact with xylene. Fits most cover-slipper instruments. Tape is 0.17 mm thick, 24.78 mm wide, and 70 m long.
Catalog No. 50-303-44