Learn More
Electron Microscopy Sciences Pre-Treated German Glass Coverslips #1, 22 mm
SDP
Catalog No. 501929534
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Sterilized by radiation, ready for your own coating. User manual included.
Diameter: 22 mm
Catalog No. 50-192-9534 Supplier Electron Microscopy Sciences Supplier No. 7229108
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Sterilized by radiation, ready for your own coating. User manual included.
Diameter: 22 mm