Learn More
Electron Microscopy Sciences CrystalView™ Coverslipping Tape 24 mm 70 MT
SDP
Catalog No. 501928621
Encompass_Preferred
CrystalView Coverslipping Film is a proprietary, resin coated film that eliminates the need for glass coverslips and sticky liquid mounting media. The flexible, tri-acetate film is coated with a xylene-activated resin on the underside, and exhibits an optical clarity comparable to glass coverslips. The coverslipped slide is free from distortion, debris, and is optimized for use on most standard automated film coverslippers. 24mm x 70 meters per roll, which can coverslip up to 1,400 slides.
Catalog No. 50-192-8621 Supplier Electron Microscopy Sciences Supplier No. 77014
CrystalView Coverslipping Film is a proprietary, resin coated film that eliminates the need for glass coverslips and sticky liquid mounting media. The flexible, tri-acetate film is coated with a xylene-activated resin on the underside, and exhibits an optical clarity comparable to glass coverslips. The coverslipped slide is free from distortion, debris, and is optimized for use on most standard automated film coverslippers. 24mm x 70 meters per roll, which can coverslip up to 1,400 slides.