Learn More
Neuvitro CE certified High precision German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1.5H thick 100 /pk untreated
Catalog No. NC1741521
Encompass_Preferred
CE certified cell culture tested High precision German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1.5H thickness 100 pieces/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.
Catalog No. NC1741521 Supplier Neuvitro Supplier No. GG1215H
CE certified cell culture tested High precision German glass coverslips for cell culture and confocal /IHC imaging, 12mm diameter #1.5H thickness 100 pieces/pk untreated. Pretreatment is required prior to cell culture applications. Shelf life 5 years, store at room temperature in dry.