Learn More
Ibidi USA Glass Coverslips for sticky-Slides (No. 1.5H, 25 mm x 75 mm, Unsterile)
SDP
Catalog No. 501149035
Encompass
Glass Coverslips for sticky-Slides. They have been designed for high–end microscopic analysis with uncompromised resolutionand choice of wavelength. No. 1.5H (170 µm +/- 5 µm) D 263 M Schott glass, 25 mm x 75 mm, unsterile, 100 pcs. /box
This item is not returnable. View return policy
Catalog No. 50-114-9035 Supplier Ibidi USA Supplier No. 10812
Glass Coverslips for sticky-Slides. They have been designed for high–end microscopic analysis with uncompromised resolutionand choice of wavelength. No. 1.5H (170 µm +/- 5 µm) D 263 M Schott glass, 25 mm x 75 mm, unsterile, 100 pcs. /box