Learn More
Ibidi USA Polymer Coverslips for sticky-Slides (No. 1.5, 25 mm x 75 mm, Unsterile, ibiTreat, Tissue-Culture Treated)
SDP
Catalog No. NC1180546
Encompass
Polymer Coverslips for sticky-Slides. They have been designed for high–end microscopic analysis with uncompromised resolutionand choice of wavelength. No. 1.5, 25 mm x 75 mm, tissue culture treated, unsterile, 100 pcs./box
This item is not returnable. View return policy
Catalog No. NC1180546 Supplier Ibidi USA Supplier No. 10814
Polymer Coverslips for sticky-Slides. They have been designed for high–end microscopic analysis with uncompromised resolutionand choice of wavelength. No. 1.5, 25 mm x 75 mm, tissue culture treated, unsterile, 100 pcs./box