Learn More
Ted Pella Inc Coverslips, Glass, 10mm dia., Thickness #1.5, 0.16-0.19mm, 100/pk
SDP
Catalog No. NC1540971
Encompass
Made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in pkg/100. Not sterile.
Catalog No. NC1540971 Supplier Ted Pella Inc Supplier No. 260368
Made from clear, high transmissive boro silicate glass. Cleaned, cut and packed in pkg/100. Not sterile.