Learn More
Electron Microscopy Sciences German Glass Coverslips #1.5, 20 mm
SDP
Catalog No. 501929525
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 20 mm
Catalog No. 50-192-9525 Supplier Electron Microscopy Sciences Supplier No. 7229011
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 20 mm