Learn More
Electron Microscopy Sciences German Glass Coverslips #1.5, 25 mm
SDP
Catalog No. 501929526
Encompass_Preferred
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 25 mm
Catalog No. 50-192-9526 Supplier Electron Microscopy Sciences Supplier No. 7229012
CE certified crystal clear German glass coverslips for cell culture and fluorescence imaging. Tested for cell and neuron cultures. Untreated (pre-treatment required before cell culture).
Diameter: 25 mm