Learn More
Research Products International Corp CoverWell Incubation Chamber, 500ul Capacity, 22 x 40mm, 50 per Package
SDP
Catalog No. 50212506
Encompass
When pressed onto a microscope slide, CoverWell forms a sealed chamber around a specimen holding reactants in place and prevents evaporation and sample loss. Designed for immunocytochemistry, in situ hybridization and enzyme/receptor cytochemistry. Convenient and easy to use. Simply press CoverWell to a microscope slide containing a cell or tissue specimen. Forms a sealed chamber and prevents evaporation, spillage, unwanted binding of antibodies, enzyme substrates, or nucleotide probes. CoverWell is made of a silicone gasket bonded to optically clear, hydrophobic plastic. Microwave safe and autoclavable.
Catalog No. 50-212-506 Supplier Research Products International Corp Supplier No. 247002
When pressed onto a microscope slide, CoverWell forms a sealed chamber around a specimen holding reactants in place and prevents evaporation and sample loss. Designed for immunocytochemistry, in situ hybridization and enzyme/receptor cytochemistry. Convenient and easy to use. Simply press CoverWell to a microscope slide containing a cell or tissue specimen. Forms a sealed chamber and prevents evaporation, spillage, unwanted binding of antibodies, enzyme substrates, or nucleotide probes. CoverWell is made of a silicone gasket bonded to optically clear, hydrophobic plastic. Microwave safe and autoclavable.