Learn More
Grace Bio Labs Inc CoverWell™ Incubation Chambers, 25/pk
Catalog No. NC1400448
Encompass_Preferred
22mm X 40mm X 0.2mm Depth, 25mm X 44mm OD, Approx. Vol. 200UL
Catalog No. NC1400448 Supplier Grace Bio Labs Inc Supplier No. 645402
22mm X 40mm X 0.2mm Depth, 25mm X 44mm OD, Approx. Vol. 200UL