Learn More
IHEALTH LABS INC iHealth COVID-19 Antigen Rapid Test (2 Pack)
SDP
Catalog No. NC3371309
Encompass_Preferred
The iHealth COVID-19 Antigen Rapid Test is a lateral flow assay intended for the qualitative detection of nucleocapsid protein antigen from SARS-CoV-2. This test is authorized for non-prescription home use with self-collected anterior nasal (nares) swab samples from individuals aged 15 years or older with symptoms of COVID-19 within the first 7 days of symptom onset. This test is also authorized for non-prescription home use with adult-collected nasal swab samples from individuals aged 2 years or older with symptoms of COVID-19 within the first 7 days of symptom onset.This test is also authorized for non-prescription home use with selfcollected anterior nasal (nares) swab samples from individuals aged 15 years or older, or adult collected anterior nasal swab samples from individuals aged 2 years or older, with or without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over 3 days with at least 24 hours (and no more than 48 hours) between tests."
Catalog No. NC3371309 Supplier IHEALTH LABS INC Supplier No. COVAG2
The iHealth COVID-19 Antigen Rapid Test is a lateral flow assay intended for the qualitative detection of nucleocapsid protein antigen from SARS-CoV-2. This test is authorized for non-prescription home use with self-collected anterior nasal (nares) swab samples from individuals aged 15 years or older with symptoms of COVID-19 within the first 7 days of symptom onset. This test is also authorized for non-prescription home use with adult-collected nasal swab samples from individuals aged 2 years or older with symptoms of COVID-19 within the first 7 days of symptom onset.This test is also authorized for non-prescription home use with selfcollected anterior nasal (nares) swab samples from individuals aged 15 years or older, or adult collected anterior nasal swab samples from individuals aged 2 years or older, with or without symptoms or other epidemiological reasons to suspect COVID-19 when tested twice over 3 days with at least 24 hours (and no more than 48 hours) between tests."