Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

InBios International, Inc. COVID-19 Detect™ Rapid Self-Test
SDP

Catalog No. 22147114
Change view
Click to view available options
Form:
Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
22-147-114 Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-147-114 Supplier InBios International, Inc. Supplier No. C19S2
Add to Cart
Add to Cart

The COVID-19 Detect™ Rapid Self-Test is a lateral flow immunochromatographic assay for the rapid, qualitative detection of the SARS-CoV-2 nucleocapsid protein antigen directly in anterior nasal swab specimens from individuals with signs and symptoms of upper respiratory tract infection.

  • Simple test requires no mixing and can be read visually
  • Sequence analysis suggests test will detect variants currently in circulation such as KP.3.1.1
  • Fast results within fifteen minutes
  • All-in-one Kit - 2 rapid tests with separately packaged swabs and pre-measured buffer tubes

Specifications

CE Marker No
Clia Complexity Waived (OTC)
Detectable Analytes COVID-19
DoA Calibrators No
Type Rapid Test
Disposable Single-use
Sterility Non-sterile
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.