Learn More
InBios International, Inc. COVID-19/Flu A and B RespiraDX™ Rapid Self-Test
Description
COVID-19/Flu A and B RespiraDX™ Rapid Self-Test is intended for the qualitative detection and differentiation of influenza A, and influenza B nucleoprotein antigens and SARS-CoV-2 nucleocapsid antigen directly in anterior nasal swab samples from individuals with signs and symptoms of respiratory tract infection.
- FDA-cleared for in vitro diagnostic use
- For over-the-counter use
- Simultaneously detects 3 viruses (SARS-CoV-2, Flu Aand B)
- No repeat testing necessary
- Two result windows for easy readability
- Shallow nasal swab collection
- Suitable for 2+ years old. Swabbing should be performed by an adult for children aged 2 to 13. User must be aged 14 + to perform self test.
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived (OTC) |
| Detectable Analytes | COVID-19 |
| DoA Calibrators | No |
| Type | Rapid Test |
| Disposable | Single-use |
| Form | Swab |