Learn More
GENABIO Covid-19 Rapid Self-Test Kit (OTC)
Description
- 1 Test/Kit
- For persons 2 years and older
- 18 month printed dating
- Supreme sensitivity against omicron variants
- Anti-counterfeit QR Code
- 252 tests/CASE
- 20 cases/Pallet (5,040 tests/pallet)
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived (EUA, OTC) |
| Detectable Analytes | SARS-CoV-2 |
| Disposable | Single-use |
| Includes | Anterioir Nasal Swab, Pre-Filled Tube, Instructions for Use, Covid-19 Test Card |
| Sterility | Sterile |
| Test Time | 15 min. |
| Form | Ready-to-Use (RTU) |