Learn More
Genabio™ Covid-19 Rapid Self-test Kit (OTC)
Description
- 2 tests/kit
- 18 month printed dating
- High sensitivity against omicron variants
- Anti-counterfeit QR Code
Specifications
Specifications
| Certifications/Compliance | FDA EUA for OTC Use |
| Clia Complexity | Not Applicable |
| Detectable Analytes | SARS-CoV-2 |
| Detection Method | Antigen |
| Format | Cassette |
| Sample Type | Nasal |
| Type | Covid Self-test |
| Disposable | Single-use |
| For Use With (Application) | Lateral Flow Rapid Test |
| Quantity | 2 Tests/Box |
| Show More |
FDA EUA for OTC.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.