Learn More
Raybiotech Inc COVID-19 Spike-ACE2 Binding Assay Kit
Catalog No. NC1816953
Encompass_Preferred
The RayBio COVID-19 Spike-ACE2 binding assay kit is a rapid, simple, and sensitive method to characterize the binding affinity of the SARS-CoV-2 Spike (S) protein and the Angiotensin I Converting Enzyme 2 (ACE2) receptor complex in the presence of potential inhibitors.
Catalog No. NC1816953 Supplier Raybiotech Inc Supplier No. COVSACE21
The RayBio COVID-19 Spike-ACE2 binding assay kit is a rapid, simple, and sensitive method to characterize the binding affinity of the SARS-CoV-2 Spike (S) protein and the Angiotensin I Converting Enzyme 2 (ACE2) receptor complex in the presence of potential inhibitors.