Learn More
Cayman Chemical COVID N1P-N2P RT-qPCR asy 1ea
SDP
Catalog No. 502945605
Encompass_Preferred
The primers and probes in the SARS CoV2 N1PN2P kit are provided in tube 1 as a 20X concentrated working solution. The same mix also contains primers/probe targeting human RPP30 mRNA splce juction as an qPCR positve control for human samples. N1P detects Omicron variants BA.1, BA.2, BA.4, and BA.5 through JN.1; N2P detects: Wuhan strain or later alpha, beta, gamma, or delta VOCs. Organism Type: Virus (+)ssRNA Control (recommended): human RPP30_RNA Enzyme Master Mixes (recommended): Empirical RT-qPCR mastermix, Standard RT-qPCR enzyme mastermix (Item No. 10706) Panels: COVID, Viral-URP Colors: CalFluorRed610 (SARS CoV-2 Delta), FAM (SARS CoV-2 Omicron), HEX (RPP30) Instruments: Any instrument
Catalog No. 50-294-5605 Supplier Cayman Chemical Supplier No. 406791 EA
The primers and probes in the SARS CoV2 N1PN2P kit are provided in tube 1 as a 20X concentrated working solution. The same mix also contains primers/probe targeting human RPP30 mRNA splce juction as an qPCR positve control for human samples. N1P detects Omicron variants BA.1, BA.2, BA.4, and BA.5 through JN.1; N2P detects: Wuhan strain or later alpha, beta, gamma, or delta VOCs. Organism Type: Virus (+)ssRNA Control (recommended): human RPP30_RNA Enzyme Master Mixes (recommended): Empirical RT-qPCR mastermix, Standard RT-qPCR enzyme mastermix (Item No. 10706) Panels: COVID, Viral-URP Colors: CalFluorRed610 (SARS CoV-2 Delta), FAM (SARS CoV-2 Omicron), HEX (RPP30) Instruments: Any instrument