Learn More
Enzo Life Sciences Coronavirus (COVID-19, MERS and SARS-CoV NP) monoclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ENZABS8061000
Encompass_Preferred
Isotype: IgG1. Species reactivity: Coronavirus. Applications: ELISA. Purity: Purified. Formulation: Liquid. In 10 mM PBS, pH 7.2 containing 0.1% sodium azide.
Catalog No. 50-201-3330