Learn More
Enzo Life Sciences Coronavirus (COVID-19 NP) antigen (aa 2-419), (recombinant) (His-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2820100
Encompass_Preferred
Alternative name: Coronavirus nucleoprotein. Purity: ≥95%. Formulation: Liquid. In 25 mM Tris, 10 mM potassium carbonate, pH 7.4. Source: Produced in E. coli. Coronavirus (COVID-19) nucleoprotein (aa 2-419) is fused to a His-tag. Long Term Storage: -20°C.
Catalog No. 50-201-4093