Learn More
Enzo Life Sciences Coronavirus (COVID-19 S1) antigen (aa 1-674), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2830100
Encompass_Preferred
Alternative name: Coronavirus spike protein. Purity: ≥95%. Formulation: Liquid. In DPBS, pH 7.4. Source: Produced in a mammalian cells expression system. Coronavirus (COVID-19) spike protein (aa 1-674) is fused to a Fc-tag. Long Term Storage: -80°C.
Catalog No. 50-201-4094