Learn More
Enzo Life Sciences Coronavirus (COVID-19 and SARS-CoV NP) monoclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ENZABS8071000
Encompass_Preferred
Isotype: IgG2b. Immunogen: Recombinant SARS-CoV (aa 1-49). Species reactivity: Coronavirus. Applications: ELISA, WB. Purity: Purified. Formulation: Liquid. In 10 mM PBS, pH 7.2 containing 0.1% sodium azide.
Catalog No. 50-201-3331