Learn More
Enzo Life Sciences Coronavirus (COVID-19, Spike) polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8270100
Encompass_Preferred
Isotype: IgG. Immunogen: Peptide corresponding to 20 amino acids near the carboxy terminus of SARS-CoV-2 (COVID-19, 2019-nCoV) Spike glycoprotein. The immunogen is located within the last 50 amino acids of SARS-CoV-2 (COVID-19, 2019-nCoV) spike protein. Species reactivity: Coronavirus. Applications: ELISA. Purity: Affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-201-3351