Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical Covidien™ Alcohol Prep Pads

Catalog No. 19898098
This item is not returnable. View return policy
Catalog No. 19-898-098 Supplier Moore Medical Supplier No. 34089

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides maximum absorbency for scrubbing and cleaning

  • Non-woven sponge preps
  • Contains 70% isopropyl alcohol
  • Sterile
  • 2-ply

Specifications

Product Type 60% Isopropyl Alcohol
For Use With (Application) Provide maximum absorbency for scrubbing and cleaning
Size Medium
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.