Learn More
Enterprise Technology Solutions Covidien Biohazard Sharps Container W/Clear Hinged Lid, 2 Gal, Red
SDP
Catalog No. 5000830970
Encompass_Preferred
Container is designed to limit accidental or intentional access to used sharp items. Hinged lid accommodates a variety of Sharps sizes. Hinged lid provides temporary closure when container is not in use and permanent closure for final disposal. Lid locks for final disposal. Empty container can be nested to save storage space.
Catalog No. 50-008-30970 Supplier Enterprise Technology Solutions Supplier No. CVDSRHL100990
Container is designed to limit accidental or intentional access to used sharp items. Hinged lid accommodates a variety of Sharps sizes. Hinged lid provides temporary closure when container is not in use and permanent closure for final disposal. Lid locks for final disposal. Empty container can be nested to save storage space.