Learn More
Enterprise Technology Solutions Covidien Biohazard Sharps Container W/Clear Lid, 3 Gallon, Red
SDP
Catalog No. 5000831714
Encompass_Preferred
Transparent Red Sharps Container features an always-open lid for convenient, quick, easy point-of-use sharps disposal. The 8-1/2" wide lid opening accommodates larger syringes. Economical horizontal drop maximizes container volume. Lid design limits accidental or intentional access to container contents. Nestable, autoclavable container allows visual monitoring and locks for final disposal.
Catalog No. 50-008-31714 Supplier Enterprise Technology Solutions Supplier No. CVDSTRT10021R
Transparent Red Sharps Container features an always-open lid for convenient, quick, easy point-of-use sharps disposal. The 8-1/2" wide lid opening accommodates larger syringes. Economical horizontal drop maximizes container volume. Lid design limits accidental or intentional access to container contents. Nestable, autoclavable container allows visual monitoring and locks for final disposal.