Learn More
Enterprise Technology Solutions Covidien Biohazard Sharps Refill w/Lift Assist Lid, 3 Gallon, Red
SDP
Supplier:  Enterprise Technology Solutions CVDS3GR100537
Encompass_Preferred
Sharps refill container is designed for use with Sharpstar In-Room Enclosure System. Design minimizes the potential for overfilling by stopping in the "FULL" position when container reaches maximum capacity. Economical horizontal drop maximizes container volume. Counterbalanced lid design limits accidental or intentional access to container contents and allows visual monitoring of the fill level. Container locks for final disposal.
Catalog No. 50-007-90555