Learn More
Enterprise Technology Solutions Covidien Biohazard Wall Cabinet for Sharp Objects, w/ Lid, 5 Quart
SDP
Catalog No. 5000790155
Encompass_Preferred
In-Room Locking Wall Mount Enclosure safely secures sharps containers and is designed to limit accidental or intentional access to used sharps. The complete In-Room System includes a 5 quart Locking Wall Mount Unit, and a 5 quart Sharpstar Transparent Red Sharps Container with lid. Easily monitor container fill levels through front and side viewing windows. Economical horizontal drop maximizes container volume. Container locks for final disposal. Neutral beige color enclosure features keyed-alike locks for convenient security. Enclosure includes container with Sharpstar lid. Enclosure is made with sturdy ABS plastic for extra protection of the sharps container and long-lasting use.
Catalog No. 50-007-90155 Supplier Enterprise Technology Solutions Supplier No. CVDSWMU100609
In-Room Locking Wall Mount Enclosure safely secures sharps containers and is designed to limit accidental or intentional access to used sharps. The complete In-Room System includes a 5 quart Locking Wall Mount Unit, and a 5 quart Sharpstar Transparent Red Sharps Container with lid. Easily monitor container fill levels through front and side viewing windows. Economical horizontal drop maximizes container volume. Container locks for final disposal. Neutral beige color enclosure features keyed-alike locks for convenient security. Enclosure includes container with Sharpstar lid. Enclosure is made with sturdy ABS plastic for extra protection of the sharps container and long-lasting use.