Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Calculi Strainer

Catalog No. 14375151
Change view
Click to view available options
Volume (Metric):
59.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-375-151 59.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-375-151 Supplier Covidien Supplier No. 2110SA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Durable, easy-to-handle strainer

  • Fits conveniently into 24-hour specimen collection containers

Specifications

Product Type Calculi Strainer
Color White
For Use With (Equipment) Specimen collection containers
Volume (English) 2 oz.
Volume (Metric) 59.1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.