Learn More
Covidien GATORGUARD™ In-Room Sharps Container
Catalog No. 22037962
Change view
Click to view available options
Capacity (English):
2 gal.
Catalog No. Capacity (English)
22-037-962 2 gal.
1 options
This item is not returnable. View return policy
Catalog No. 22-037-962 Supplier Covidien Supplier No. 31323333
Add to Cart
Add to Cart
This item is not returnable. View return policy
Efficient disposal with counterbalance door
  • Counterbalance door design allows for the efficient disposal of devices of varying sizes, from a lightweight sharps to a 60cc syringe
  • Recommended fill line is easily visible
  • “FULL” indicator signals when the container capacity has been reached
  • Translucent Red
Specifications
Capacity (Metric) 7.6 L
Closure Type Counter-Balanced Door
Capacity (English) 2 gal.
Color Translucent Red
Material Plastic