Learn More
Leica Microsystems Covidien Microhematocrit Tube Reader
Catalog No. 22260984
Encompass_Preferred
Change view
Click to view available options
Product Type:
Microhematocrit Tube Reader
Catalog No. Product Type
22-260984 Microhematocrit Tube Reader
1 options
Catalog No. 22-260984 Supplier Leica Microsystems Supplier No. 39111004
To be used with any sealed or capped 75mm microhematocrit tube
  • Handy, pocket-sized, universal tube reader has two ruled charts for precise determination of microhematocrit values
  • Made of durable, washable, vinyl-laminated plastic
TRUSTED_SUSTAINABILITY
Specifications
Size Universal, pocket sized
Product Type Microhematocrit Tube Reader
For Use With (Equipment) Any sealed or capped 75mm microhematocrit tube
Includes 2 Ruled charts
Material Vinyl-laminated Plastic