Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Midstream Collection with Preservative

Catalog No. 14375191
Change view
Click to view available options
Includes:
Preservative
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
14-375-191 Preservative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-375-191 Supplier Covidien Supplier No. 7000SA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Leak-resistant systems maintain specimen integrity for up to 24 hours

  • Effective for transport in pneumatic tube systems
  • Packaging not only guides the patient through procedure, but also allows patient to view instructions
  • Metal twist cap minimizes leakage with unique Plastisol™ lining

Specifications

Product Type Urine Specimen System
For Use With (Application) Leak-resistant systems maintain specimen integrity for up to 24 Hours
Includes Preservative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.