Learn More
Covidien Monoject™ Angel Wing™ Blood Collection/Infusion Sets with Female Luer
Catalog No. 22257424
Change view
Click to view available options
Needle Gauge:
19 G
21 G
23 G
25 G
Color:
Blue
Cream
Green
Orange
Length (English) Tube:
3 in.
12 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color Length (English) Tube
22-257-424 23 G Blue 3 in.
22-225174 19 G Cream 12 in.
22-225182 21 G Green 12 in.
22-042907 23 G Blue 12 in.
05-561-45 25 G Orange 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-257-424 Supplier Covidien Supplier No. 8881225390
This item is not returnable. View return policy
Used for routine venipunctures and infusion applications
  • One-handed activation of safety mechanism prevents accidental needlesticks
  • Textured wings identify bevel and provide ease of control
  • Flexible tubing prevents coiling
  • Latex-free to minimize potential allergic reaction
  • 0.75 in. (1.9cm) needle length
Specifications
Product Type Winged Blood Collection Set
Color Blue
Needle Gauge 23 G
Length (English) Needle 0.75 in.
Length (English) Tube 3 in.