Learn More
Covidien Monoject™ Angel Wing™ Blood Collection/Infusion Sets with Multi-Sample Luer Adapter
Catalog No. 22225307
Change view
Click to view available options
Needle Gauge:
19 G
21 G
23 G
25 G
Color:
Blue
Cream
Green
Orange
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-225307 23 G Blue
22-225-281 19 G Cream
05-561-46 25 G Orange
22-225299 21 G Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-225307 Supplier Covidien Supplier No. 8881225307
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sets with female Luer attachments for use in routine venipunctures and infusion applications
  • One-handed activation of safety mechanism prevents accidental needlesticks
  • Textured wings identify bevel and provide ease of control
  • Flexible tubing prevents coiling
  • Latex-free to minimize potential allergic reaction
Specifications
Product Type Blood Collection Set
Color Blue
Needle Gauge 23 G
Includes Multi-sample Female Luer Adapter
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.