Learn More
Covidien Monoject™ Angel Wing™ Multi-Sample Female Transfer Set
Catalog No. 22031353
Change view
Click to view available options
For Use With (Application):
Transfer of blood samples
Catalog No. For Use With (Application)
22-031-353 Transfer of blood samples
1 options
This item is not returnable. View return policy
Catalog No. 22-031-353 Supplier Covidien Supplier No. 8881225241
This item is not returnable. View return policy
Designed for the safe transfer of blood from syringes directly into blood culture bottles
  • Adapter insert enables the safe collection of blood from syringes into evacuated blood tubes
  • Sterile, single use
Specifications
Product Type Multi-sample Female Transfer Set
Sterility Sterile
For Use With (Application) Transfer of blood samples