Learn More
Covidien Monoject™ Angel Wing™ Safety Female Transfer Sets
Catalog No. 22225232
Change view
Click to view available options
For Use With (Application):
Blood Transfer
Product Type:
Dome
Tube Holder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Product Type
22-225232 Blood Transfer Tube Holder
22-029-330 Blood Transfer Dome
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-225232 Supplier Covidien Supplier No. 8881225232
Safely transfer blood specimens from syringes into blood collection tubes and blood culture bottles
  • Recessed multi-sample needle protected by a plastic shield—easy application of blood collection tube
  • Sets feature needle-shielded holders for transferring blood specimens from syringes
  • Sterile
Cat. No. 22-225-232 Tube Holder is designed for transferring blood from syringes into blood collection tubes; 22-029-330 Dome for transferring blood from syringes into blood culture bottles and blood collection tubes.
Specifications
Product Type Tube Holder
Sterility Sterile
For Use With (Application) Blood Transfer