Learn More
Covidien Monoject™ Angel Wing™ Safety Male Collection Sets
Catalog No. 22225216
Change view
Click to view available options
Product Type:
Tube Holder
Catalog No. Product Type
22-225216 Tube Holder
1 options
This item is not returnable. View return policy
Catalog No. 22-225216 Supplier Covidien Supplier No. 8881225216
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for use with Angel Wing blood collection sets
  • Safely transfer blood specimens from syringes into blood collection tubes and blood culture bottles
  • Recessed multi-sample needle protected by a plastic shield—easy application of blood collection tube
  • Sets feature needle-shielded holders for blood collection
  • Sterile
Cat. No. 22-225-216 Tube Holder is designed for use with evacuated blood collection tubes; 22-225-224 Dome for use with blood culture and blood collection tubes.
Specifications
Product Type Tube Holder
Sterility Sterile
Includes Holder and Safety Cap