Learn More
Covidien Monoject™ Blood Collection Device
Catalog No. 22031354
Change view
Click to view available options
Product Type:
Male Blood Collection Set
Catalog No. Product Type
22-031-354 Male Blood Collection Set
1 options
This item is not returnable. View return policy
Catalog No. 22-031-354 Supplier Covidien Supplier No. 8881225245
This item is not returnable. View return policy
Requires
BacT/Alert blood culture bottles
Designed to specifically fit BacT/Alert™ blood culture bottles
  • Single use for safe transfer of blood samples and reduction of needlestick injuries
  • Sterile to help reduce false positive readings
  • Adapter insert for evacuated tubes
  • Male
Specifications
Product Type Male Blood Collection Set
Sterility Sterile
For Use With (Application) Blood collection