Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Blood Collection/Infusion Sets with Multi-Sample Luer Adapter

Catalog No. 22257426
Change view
Click to view available options
Needle Gauge:
23 G
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-257-426 23 G Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-257-426 Supplier Covidien Supplier No. 8881225707

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Product Type Blood Collection Set
Color Blue
Needle Gauge 23 G
For Use With (Application) Enipunctures and infusion applications
Includes MSLA
Length (English) Needle 0.75 in.
Length (English) Tube 7 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.