Learn More
Covidien Monoject™ Bone Marrow Biopsy Trays with Safety Components
Description
- Rigid stainless-steel needle features a double-cutting edge cannula and sharp style for confident use and smoother entry into the marrow
- Comfortable ergonomic handle design provides precision control with minimal discomfort
- Conveniently packaged in sterile procedural trays that comply with industry standards
Compliance
In compliance with the current FDA Quality System Requirements (QSR) as stipulated in 21 CFR Part 820.
Specifications
Specifications
| Needle Gauge | 11 G |
| For Use With (Application) | For bone marrow collection |
| Includes | Safety components, 11G bone marrow needle |
| Product Type | Biopsy Tray |