Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Silicone-Coated Tubes with Red Stoppers

Catalog No. 22029324
Change view
Click to view available options
Capacity (Metric):
5 mL
7 mL
10 mL
15 mL
Tube Size:
13 x 75 mm
13 x 100 mm
16 x 100 mm
16 x 125 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Tube Size
22-029324 5 mL 13 x 75 mm
22-301710 10 mL 16 x 100 mm
22-029316 15 mL 16 x 125 mm
22-032264 7 mL 13 x 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-029324 Supplier Covidien Supplier No. 8881301413
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Standard blood-collection tubes

  • Interior coated with water-soluble silicone material
  • Sterilization applies to the tube and stopper interior only, and not to the outside of the tube or to the packaging
  • Sterilized by gamma radiation
  • Used for laboratory procedures requiring serum; general chemistry and serology procedures

Specifications

Product Type Silicone Coated Tube
Additive No Additive
Sterility Sterile
Capacity (Metric) 5 mL
Closure Color Red
Tube Size 13 x 75 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.