Learn More
Covidien Monoject™ SoftPack Tuberculin Syringes
Catalog No. 22257150
Change view
Click to view available options
Needle Gauge:
25 G
26 G
27 G
28 G
No Needle Included
Connection Type:
Detachable
Fixed
Luer
Luer Tip
Removable
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Connection Type
22-257-150 No Needle Included Luer Tip
22-257-135 No Needle Included Luer
22-257-136 25 G Removable
22-257-138 26 G Detachable
22-257-139 27 G Detachable
22-257-137 25 G Fixed
22-257-140 28 G Fixed
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-257-150 Supplier Covidien Supplier No. 1180100777
Add to Cart
Add to Cart
Sterile, single-use individually packaged syringes
  • Polypropylene barrel and plunger rod
  • Latex-free plunger tip
  • Graduated barrel markings
  • Variety of needle gauge sizes
Specifications
Material Polypropylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Softpack Tuberculin Syringe
Disposable Single-use
Graduations 0.01 mL
Volume (Metric) 1 mL
Connection Type Luer Tip
Graduated Yes
Sterility Sterile