Learn More
Enterprise Technology Solutions Covidien Phlebotomy Sharps Container W/Clear Lid, Flip Top,1 Qt.,RD
SDP
Catalog No. 5000830225
Encompass_Preferred
Container is designed to limit accidental or intentional access to used sharp items. Empty containers can be nested to save storage space. Hinged lid provides temporary closure when container is not in use and permanent closure for final disposal. Flip-top lid locks for final disposal. Use of wall brackets (sold separately) frees up valuable counter space.
Catalog No. 50-008-30225 Supplier Enterprise Technology Solutions Supplier No. CVDSR1Q100900
Container is designed to limit accidental or intentional access to used sharp items. Empty containers can be nested to save storage space. Hinged lid provides temporary closure when container is not in use and permanent closure for final disposal. Flip-top lid locks for final disposal. Use of wall brackets (sold separately) frees up valuable counter space.