Learn More
Covidien Precision™ 24-Hour Specimen Containers
Catalog No. p-180797
Change view
Click to view available options
Volume (Metric):
3000 mL
Closure Type:
Pour-spout Cap
Screw Cap
No. per Case:
40/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Closure Type No. per Case
14-375-186 3000 mL Pour-spout Cap 40/Cs.
14-375-185 3000 mL Screw Cap 40/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-375-186 Supplier Covidien Supplier No. 5010SA
This item is not returnable. View return policy
Simplify urine output collection process by providing easy access, leak resistance, and protection from light
  • Convenient pouring spout for quick, safe transfer of urine sample
  • Spout is self-venting when pouring
Specifications
Product Type 24-Hour Urine Specimen Collection Container
Closure Type Pour-spout Cap
Graduated Yes
Closure Material Plastic
Color Amber
Includes Pour spout
No. per Case 40/Cs.
Volume (Metric) 3000 mL