Learn More
Covidien Precision™ OR Sterile Specimen Container
Catalog No. 14375181
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
14-375-181 Sterile
1 options
Catalog No. 14-375-181 Supplier Covidien Supplier No. 2600SA
Only null left
Add to Cart
Add to Cart
Sterile specimen containers with tamper-evident seals for leak-resistant transport in pneumatic tube systems
  • Clear plastic construction with graduations offers easy observation and measurement of contents
  • Highly visible “Positive Seal” indicator clearly shows container is sealed when red bars are lined up
  • Exclusive metal lid with plastic lining provides a leak-resistant seal to minimize spillage of specimen during transportation
Specifications
Graduated Yes
Material Plastic
Sterility Sterile
Capacity (English) 4 oz.
Closure Material Metal
Closure Type Positive Seal Indicator
Color Clear