Learn More
Enterprise Technology Solutions Covidien Sharps Container System,Locking,Glovebox,Wall Mount, 5 Qt.
SDP
Catalog No. 5000790916
Encompass_Preferred
Complete In-Room Wall Mount Enclosure safely secures Sharps containers and is designed to limit accidental or intentional access to used sharps. The system includes a 5 quart Locking Wall Mount Unit, a 5 quart Sharpstar Transparent Red Sharps Container with lid, and one Glove Dispenser. Easily monitor container fill levels through front and side-viewing windows. Economical horizontal drop maximizes container volume. Container locks for final disposal. Neutral beige color enclosure features keyed-alike locks for convenient security. Enclosure is made with sturdy ABS plastic for extra protection of the Sharps container and long-lasting use.
Catalog No. 50-007-90916 Supplier Enterprise Technology Solutions Supplier No. CVDSSGB10056H
Complete In-Room Wall Mount Enclosure safely secures Sharps containers and is designed to limit accidental or intentional access to used sharps. The system includes a 5 quart Locking Wall Mount Unit, a 5 quart Sharpstar Transparent Red Sharps Container with lid, and one Glove Dispenser. Easily monitor container fill levels through front and side-viewing windows. Economical horizontal drop maximizes container volume. Container locks for final disposal. Neutral beige color enclosure features keyed-alike locks for convenient security. Enclosure is made with sturdy ABS plastic for extra protection of the Sharps container and long-lasting use.