Learn More
Enterprise Technology Solutions Covidien Sharps Container,w/ Counter Balanced Lid, 5 Quart, Red
SDP
Catalog No. 5000790382
Encompass_Preferred
Sharps refill container with counterbalanced lid is designed for use with Sharpstar In-Room Enclosure System. Design minimizes the potential for overfilling by stopping in the "FULL" position when container reaches maximum capacity. Economical horizontal drop maximizes container volume. Lid design limits accidental or intentional access to container contents and allows visual monitoring of the fill level. Container locks for final disposal.
Catalog No. 50-007-90382 Supplier Enterprise Technology Solutions Supplier No. CVDK5SS1007SA
Sharps refill container with counterbalanced lid is designed for use with Sharpstar In-Room Enclosure System. Design minimizes the potential for overfilling by stopping in the "FULL" position when container reaches maximum capacity. Economical horizontal drop maximizes container volume. Lid design limits accidental or intentional access to container contents and allows visual monitoring of the fill level. Container locks for final disposal.