Learn More
Covidien SharpSafety™ Universal Waste Containers
Catalog No. 22037969
Change view
Click to view available options
Capacity (English):
1 qt.
2 gal.
8 gal.
Closure Type:
Counter-Balanced Lid
Hinged Top
Hinged, Snap
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Closure Type
22-037-969 2 gal. Hinged, Snap
22-037-967 1 qt. Counter-Balanced Lid
22-037-968 8 gal. Hinged Top
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-037-969 Supplier Covidien Supplier No. 8602U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to segregate pharmaceutical waste
Containers lock for final disposal to secure contents.
  • Nestable, saving valuable storage space
  • Counterbalance or hinged lids to accommodate various sizes of sharps
Specifications
Capacity (Metric) 7.57 L
Closure Type Hinged, Snap
Capacity (English) 2 gal.
For Use With (Application) Designed to segregate pharmaceutical waste