Learn More
Med Vet International McKesson Transducer Cover, 2 X 7-1/2 Inch, 16-1408, 100/Box
Catalog No. 503042923
Encompass_Preferred
Ultrasound Transducer Cover McKesson 2 X 7-1/2 Inch Latex NonSterile For use with Ultrasound External Probe 2 Inches by 7½ Inches Intended to be used as a protective transducer probe equipment covering, such as to isolate equipment from microbial and other contamination Designed specifically for ultrasound use to be used as a protective covering over ultrasound scanhead instruments and cord offering a sterile solution for a wide variety of transducers Rolled covers with a 7.5-inch length and 1.4-inch diameter to securely cover probe using aseptic technique Beaded cuff for more secure positioning Nonsterile, latex devices with magnesium carbonate powder for single use Conveniently packaged in resealable bag
Catalog No. 50-304-2923 Supplier Med Vet International Supplier No. 1087740BX
Ultrasound Transducer Cover McKesson 2 X 7-1/2 Inch Latex NonSterile For use with Ultrasound External Probe 2 Inches by 7½ Inches Intended to be used as a protective transducer probe equipment covering, such as to isolate equipment from microbial and other contamination Designed specifically for ultrasound use to be used as a protective covering over ultrasound scanhead instruments and cord offering a sterile solution for a wide variety of transducers Rolled covers with a 7.5-inch length and 1.4-inch diameter to securely cover probe using aseptic technique Beaded cuff for more secure positioning Nonsterile, latex devices with magnesium carbonate powder for single use Conveniently packaged in resealable bag